missing translation for 'onlineSavingsMsg'
Learn More

ANKRD36 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

415.00€ - 624.00€

Specifications

Antigen ANKRD36
Applications Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18206304
View Documents
Novus Biologicals
NBP2-56995
100 μL
624.00€
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18694649
View Documents
Novus Biologicals
NBP2-56995-25ul
25 μL
415.00€
25µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

ANKRD36 Polyclonal specifically detects ANKRD36 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

ANKRD36
Polyclonal
Rabbit
Human
ankyrin repeat domain 36, ankyrin repeat domain-containing protein 36, UNQ2430
ANKRD36
IgG
Affinity Purified
Immunocytochemistry, Immunofluorescence
Unconjugated
RUO
PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide
375248
This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:NGMQDSRKKLDQMRSQFQEIQDQLTATIRCTKEMEGD
Primary
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.