missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ANKRD32 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-76546-25ul
This item is not returnable.
View return policy
Description
ANKRD32 Polyclonal specifically detects ANKRD32 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
Specifications
| ANKRD32 | |
| Polyclonal | |
| Immunocytochemistry/Immunofluorescence 1-4 μg/mL | |
| ankyrin repeat domain 32, ankyrin repeat domain-containing protein 32, BRCT domain containing 1, BRCT domain-containing protein 1, BRCTD1, BRCTx, DKFZp564°C0469, DKFZp761C121 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 84250.0 | |
| Human | |
| Purified |
| Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% sodium azide | |
| ANKRD32 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: SWLQMFVAEAVFKKLCLQSSGSVSSEPLSLQKMVYSYLPALGKTGVLGSGKIQVSKKIGQRPCFDSQRTLLMLNGTKQKQVEGLP | |
| 25 μL | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction