Learn More
Novus Biologicals™ Angiopoietin-2 Antibody (Roche patent anti-ANG-2), Novus Biologicals™
Human Monoclonal Antibody
Brand:  Novus Biologicals™ NBP3-27955-50ug
 View more versions of this product
Product Code. 30045414
  • 213.00€ / 50µg
Description
Angiopoietin-2 Monoclonal antibody specifically detects Angiopoietin-2 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Angiopoietin-2
Monoclonal
Unconjugated
O15123
Human
Protein A purified
RUO
Primary
Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex.
Store at -20°C in powder form. Store at -80°C once reconstituted.
IgG1
Flow Cytometry, ELISA, Functional Assay
Roche patent anti-ANG-2
Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2)
AGPT2, ANG2, ANG-2, angiopoietin 2, angiopoietin-2, angiopoietin-2a, angiopoietin-2B, angiopoitin 2, Tie2-ligand
ANGPT2
50 μg
Angiogenesis, Cancer
285