missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Angiopoietin-2 Antibody (5A9), Novus Biologicals™
Human Monoclonal Antibody
343.00€ - 533.00€
Specifications
| Antigen | Angiopoietin-2 |
|---|---|
| Clone | 5A9 |
| Dilution | ELISA |
| Applications | ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30043654
|
Novus Biologicals
NBP3-26508-50ul |
50 μL |
343.00€
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30043102
|
Novus Biologicals
NBP3-26508-100ul |
100 μL |
533.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Angiopoietin-2 Monoclonal antibody specifically detects Angiopoietin-2 in Human samples. It is validated for ELISASpecifications
| Angiopoietin-2 | |
| ELISA | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| AGPT2, ANG2, ANG-2, angiopoietin 2, angiopoietin-2, angiopoietin-2a, angiopoietin-2B, angiopoitin 2, Tie2-ligand | |
| Recombinant Human Angiopoietin-2 protein [UniProt O15123] | |
| Primary | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. |
| 5A9 | |
| ELISA | |
| Unconjugated | |
| Human | |
| Angiogenesis, Cancer | |
| 50% Glycerol, 0.01M PBS, PH 7.4 | |
| 285 | |
| IgG1 | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title