Learn More
Invitrogen™ Angiogenin Polyclonal Antibody
Goat Polyclonal Antibody
Brand: Invitrogen™ PA5148449
This item is not returnable.
View return policy
Description
Reconstitute at 1 mg/mL in sterile PBS.
Angiogenin (ANG or ANG I) is important for the process of neovascularization and formation of new blood vessels. ANG is similar to pancreatic ribonuclease A and functions as a tRNA-specific ribonuclease that abolishes protein synthesis by specifically hydrolyzing cellular tRNAs. It interacts with endothelial cell-surface actin and may cause changes in the cell cytoskeleton. ANG is thought to be involved in the development of solid tumors and its antagonists are capable of inhibiting tumor growth. Defects in ANG are the cause of susceptibility to amyotrophic lateral sclerosis type 9 (ALS9). Angiogenin is a genetic link between ALS and PD.
Specifications
| Angiogenin | |
| Polyclonal | |
| Unconjugated | |
| ANG | |
| AI385586; ALS9; ANG; Ang1; Angiogenin; angiogenin ribonuclease 1; angiogenin, ribonuclease A family, member 1; angiogenin, ribonuclease, RNase A family, 5; angiogenin-1; epididymis luminal protein 168; HEL168; MGC22466; MGC71966; RAA1; Ribonuclease 5; ribonuclease A A1; ribonuclease A family member 5; RNase 5; RNASE4; RNASE5; Rnase5a | |
| Goat | |
| Protein A/G | |
| RUO | |
| 283 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
| Neutralization, Western Blot | |
| 1 mg/mL | |
| PBS with trehalose and no preservative | |
| P03950 | |
| ANG | |
| E. coli-derived recombinant human Angiogenin, Gln25-Pro147. | |
| 1 mg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |