missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ANAPC13 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
241.00€ - 326.00€
Specifications
| Antigen | ANAPC13 |
|---|---|
| Dilution | Immunohistochemistry-Paraffin 1:50-1:200 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18613172
|
Novus Biologicals
NBP2-97529-100ul |
100 μg |
326.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18675934
|
Novus Biologicals
NBP2-97529-50ul |
50 μg |
241.00€
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ANAPC13 Polyclonal antibody specifically detects ANAPC13 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
| ANAPC13 | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Cell Biology | |
| PBS (pH 7.0) | |
| 25847 | |
| IgG | |
| Antigen and protein A Affinity-purified |
| Immunohistochemistry-Paraffin 1:50-1:200 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| anaphase promoting complex subunit 13, anaphase-promoting complex subunit 13, APC13SWM1, cyclosome subunit 13, Cylosome subunit 13, DKFZP566D193, homolog of yeast Swm1 | |
| Produced in rabbits immunized with E. coli-derived Human ANAPC13 fragment. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title