missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ Amyloid A4/Amyloid β Ab-1, Mouse Monoclonal Antibody

Obtain accurate, reproducible results in immunohistochemistry experiments with Epredia™ Amyloid A4/Amyloid β Ab-1, Mouse Monoclonal Antibody.

Brand:  Epredia™ MS-1167-S1

 View more versions of this product

Product Code. 12649176

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products »

This item is not returnable. View return policy

Description

Description

Amyloid beta-protein/amyloid A4 or amyloid beta-peptide is derived from the amyloid beta-precursor protein. Beta amyloid is an extracellular filamentous protein deposit found in the brain. It is the major protein component of amyloid cores, neuritic plaques and is also found as a deposit in neurofibrillary tangles. In humans, Alzheimer's disease is the most common cause of senile dementia and is characterized by abnormal filamentous protein deposits in the brain. Beta amyloid deposits are also detected in Lewy body dementia, Down's syndrome, amyloidosis (Dutch type) and in the Guam Parkinson-Dementia complex.

Host Species: Mouse

Clone: BAM01; same as 6F/3D

Isotype: IgG1/κ

Species Reactivity: Human. Others not tested.

Epitope: aa 8-17 (SGYEVHHQKL) of mature amyloid A4/ amyloid Beta which is the same as aa 660-669 of human amyloid A4/amyloid β protein precursor

Immunogen: A synthetic peptide consisting of residues 8-17 (SGYEVHHQKL) of the amyloid β-protein/amyloid A4 which is the same as aa 660-669 of human amyloid A4/amyloid β protein precursor

Molecular Weight: 102kDa (precursor), 4-5kDa (mature)

Positive Control: Human brain of Alzheimer's disease

Cellular Localization: Transmembrane (APP) and extracellular (Amyloid β)

Recommended for:

  • Immunohistochemistry (Formalin/paraffin only)

Note:

USA: RUO; Int'l: RUO

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Amyloid A4/Amyloid beta Ab-1
Monoclonal
Unconjugated
A synthetic peptide consisting of residues 8-17 (SGYEVHHQKL) of the amyloid beta-protein/amyloid A4 which is the same as aa 660-669 of human amyloid A4 / amyloid Beta protein precursor
500 μL
Neuroscience
Human
IgG2a κ
Immunohistochemistry (Paraffin)
BAM01; same as 6F/3D
Mouse
Human brain of Alzheimer's disease
RUO
Primary
Concentrated
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.