missing translation for 'onlineSavingsMsg'
Learn More

Amoxicillin Dimer tri-Sodium Salt (penicilloic acid form) (Mixture of Diastereomers), TRC

Product Code. 30138220
Change view
Click to view available options
Quantity:
0.5 mg
2.5 mg
10 mg
Unit Size:
0.5mg
10mg
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30138220 2.5 mg 2.5mg
30136518 0.5 mg 0.5mg
30012395 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138220 Supplier Toronto Research Chemicals Supplier No. TRCA6345202.5MG
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Weight (g/mol) 814.77

Specifications

Analyte or Component Names Amoxicillin Dimer tri-Sodium Salt (penicilloic acid form) (Mixture of Diastereomers)
Shipping Condition Room Temperature
Molecular Formula C32H37N6Na3O11S2
Synonym Amoxicillin Dimer tri-Sodium Salt (penicilloic acid form)(Mixture of Diastereomers)
InChI Formula InChI=1S/C32H40N6O11S2.3Na/c1-31(2)21(29(46)47)37-26(50-31)19(35-23(41)17(33)13-5-9-15(39)10-6-13)25(43)34-18(14-7-11-16(40)12-8-14)24(42)36-20(28(44)45)27-38-22(30(48)49)32(3,4)51-27;;;/h5-12,17-22,26-27,37-40H,33H2,1-4H3,(H,34,43)(H,35,41)(H,36,42)(H,44,45)(H,46,47)(H,48,49);;;/q;3*+1/p-3/t17?,18-,19?,20+,21+,22+,26-,27-;;;/m1.../s1
Solution Type Neat
SMILES O=C(C(C1=CC=C(C=C1)O)N)NC([C@@]2(N[C@H](C(C)(S2)C)C(O[Na])=O)[H])C(N[C@@H](C(N[C@]([H])(C(O[Na])=O)[C@]3([H])N[C@H](C(C)(S3)C)C(O[Na])=O)=O)C4=CC=C(C=C4)O)=O
IUPAC Name sodium (2R,4S)-2-(1-(2-amino-2-(4-hydroxyphenyl)acetamido)-2-(((R)-2-(((R)-carboxylato((2R,4S)-4-carboxylato-5,5-dimethylthiazolidin-2-yl)methyl)amino)-1-(4-hydroxyphenyl)-2-oxoethyl)amino)-2-oxoethyl)-5,5-dimethylthiazolidine-4-carboxylate
Industry Type Forensics
Quantity 2.5 mg
Formula Weight 814.17
Chemical Name or Material Amoxicillin Dimer tri-Sodium Salt (penicilloic acid form) (Mixture of Diastereomers)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.