missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Amoxicillin Dimer tri-Sodium Salt (penicilloic acid form) (Mixture of Diastereomers), TRC
Specifications
Specifications
| Analyte or Component Names | Amoxicillin Dimer tri-Sodium Salt (penicilloic acid form)(Mixture of Diastereomers) |
| Shipping Condition | Room Temperature |
| Molecular Formula | C32 H37 N6 Na3 O11 S2 |
| Synonym | Amoxicillin Dimer tri-Sodium Salt (penicilloic acid form)(Mixture of Diastereomers), (2R)-(9CI)(2R)-2-(4-Hydroxyphenyl)glycyl-(2R)-2-[(2R,4S)-4-carboxy-5,5-dimethyl-2-thiazolidinyl]glycyl-(2R)-2-(4-hydroxyphenyl)glycyl-2-[(2R,4S)-4-carboxy-5,5-dimethyl-2-thiazolidinyl]glycine tri-Sodium Salt; |
| InChI Formula | InChI=1S/C32H40N6O11S2.3Na/c1-31(2)21(29(46)47)37-26(50-31)19(35-23(41)17(33)13-5-9-15(39)10-6-13)25(43)34-18(14-7-11-16(40)12-8-14)24(42)36-20(28(44)45)27-38-22(30(48)49)32(3,4)51-27;;;/h5-12,17-22,26-27,37-40H,33H2,1-4H3,(H,34,43)(H,35,41)(H,36,42)(H,44,45)(H,46,47)(H,48,49);;;/q;3*+1/p-3/t17?,18-,19?,20+,21+,22+,26-,27-;;;/m1.../s1 |
| Solution Type | Neat |
| SMILES | O=C(NC(C(N[C@H](C1=CC=C(O)C=C1)C(N[C@]([C@@]2(N[C@@H](C(O[Na])=O)C(C)(C)S2)[H])([H])C(O[Na])=O)=O)=O)[C@]3([H])N[C@@H](C(O[Na])=O)C(C)(C)S3)C(N)C4=CC=C(O)C=C4 |
| IUPAC Name | trisodium;(2R,4S)-2-[1-[[2-amino-2-(4-hydroxyphenyl)acetyl]amino]-2-[[(1R)-2-[[(R)-carboxylato-[(2R,4S)-4-carboxylato-5,5-dimethyl-1,3-thiazolidin-2-yl]methyl]amino]-1-(4-hydroxyphenyl)-2-oxoethyl]amino]-2-oxoethyl]-5,5-dimethyl-1,3-thiazolidine-4-carboxylate |
| Industry Type | Pharmaceutical |
| Quantity | 0.5 mg |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?