missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Amnionless/AMN Polyclonal antibody specifically detects Amnionless/AMN in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antigen | Amnionless/AMN |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Immunohistochemistry-Paraffin 1:50-1:200 |
| Formulation | PBS (pH 7.0) |
| Gene Alias | amnionless homolog (mouse), PRO1028, protein amnionless, UNQ513/PRO1028, visceral endoderm-specific type 1 transmembrane protein |
| Host Species | Rabbit |
| Immunogen | Produced in rabbits immunized with E. coli-derived Human Amnionless/AMN fragment. |
| Purification Method | Antigen and protein A Affinity-purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?