missing translation for 'onlineSavingsMsg'
Learn More

Ammonium zinc sulfate hydrate

Product Code. 11410114
Click to view available options
Quantity:
250 g
1000 g
5000 g
Unit Size:
1000g
250g
5000g
This item is not returnable. View return policy

Product Code. 11410114

Brand: Thermo Scientific Alfa Aesar A13584.30

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 77998-33-5 | H8N2O8S2Zn | 293.57 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 77998-33-5
Molecular Formula H8N2O8S2Zn
Molecular Weight (g/mol) 293.57
MDL Number MFCD18711942
InChI Key CHORECZHRAUKSW-UHFFFAOYSA-L
Synonym ammonium zinc sulfate hydrate, acmc-20alsa, ammonium hydrate zinc sulfate, cvgyqralnwebmp-uhfffaoysa-m, ammonium ? 1-zinc 1+ hydrate sulfate, sulfuric acid, ammoniumzinc salt 2:2:1 , hydrate 9ci
PubChem CID 53471899
SMILES [NH4+].[NH4+].[Zn++].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O

Specifications

Quantity 250 g
UN Number UN3077
IUPAC Name azanium;zinc;sulfate;hydrate
Formula Weight 293.57 (Anhydrous)
Density 1.931
Chemical Name or Material Ammonium zinc sulfate hydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.