accessibility menu, dialog, popup

UserName

CAS RN 77998-33-5

NH4OSOOOZnOSOOONH4

CAS RN 77998-33-5

IUPAC Name: ammonium hydrate zinc sulfate
Synonyms: ammonium hydrate zinc sulfate
Molecular Weight (g/mol): 197.49
Molecular Formula: H6NO5SZn
InChi Key: GMRYSHZVWPTLCH-UHFFFAOYSA-M
SMILES: [NH4+].O.[Zn].[O-]S([O-])(=O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)