missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium Chloride Buffer Solution, According to European Pharmacopoeia (EP) Chapter 4 (4.1.3), pH 9.5, Reagecon
Shop All Reagecon Products

Click to view available options
Quantity:
1 L
Unit Size:
1 L
Description
- Tested by ISO 17025 accredited test method
Specifications
Specifications
| Chemical Name or Material | Buffer solution |
| For Use With (Application) | European Pharmacopoeia monograms |
| pH | 9.5 |
| Physical Form | Liquid |
| Quantity | 1 L |
| Packaging | HDPE bottle GL45 |
| UN Number | 1760 |
| Traceability to NIST | Yes |
Safety and Handling
Product Identifier
- ammonium chloride buffer solution pH9.5
- Danger
- Skin corrosion/irritation Category 1 B
- Serious eye damage/eye irritation Category 1
- Specific target organ toxicity Category 3
- H314-Causes severe skin burns and eye damage.
- H335-May cause respiratory irritation.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P321-Specific treatment (see on this label).
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : ammonia, ammonium chloride
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction