missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Alkylbenzenesulfonate (Mixture of different chain lengths), TRC

Product Code. 30335858
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30335858 10 g 10g
30331932 5 g 5g
30336756 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30335858 Supplier Toronto Research Chemicals Supplier No. TRCA63405510G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ammonium Alkylbenzenesulfonate (Mixture of different chain lengths)
Analyte or Component Names Ammonium Alkylbenzenesulfonate (Mixture of different chain lengths)
Shipping Condition Room Temperature
Solution Type Neat
SMILES O=S(C1=CC=CC=C1)(O)=O.CCCCCCCCCCCCC.N
Industry Type Pharmaceutical
Quantity 10 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.