missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Acetate, TRC

Product Code. 17987934
Change view
Click to view available options
Quantity:
10 g
50 g
100 g
Unit Size:
100g
10g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17987934 10 g 10g
17927194 100 g 100g
17938034 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17987934 Supplier Toronto Research Chemicals Supplier No. TRCA63390010G
 Restricted Item
Estimated Shipment: 03-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 631-61-8
Molecular Formula C2 H3 O2 . H4 N
Molecular Weight (g/mol) 77.08
Synonym Acetic Acid Ammonium Salt, Mindererus's Spirit, 1BEEM;
IUPAC Name azanium;acetate
SMILES [NH4+].CC(=O)[O-]

Specifications

Analyte or Component Names Ammonium acetate
Shipping Condition Room Temperature
InChI Formula InChI=1S/C2H4O2.H3N/c1-2(3)4;/h1H3,(H,3,4);1H3
Solution Type Neat
Industry Type Forensics
Quantity 10 g
Formula Weight 77.0477
Chemical Name or Material Ammonium acetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.