Learn More
Ammonia Dilute R3 solution EU Pharmacopoeia, Fisher Chemical™
Shop All Fisher Chemical Products
Click to view available options
Quantity:
1000 mL
Unit Size:
1000mL
Specifications
Specifications
| Packaging | Bottle |
| Analyte or Component Names | Ammonia, dilute R3 |
| Purity Grade Notes | EU Ph Ref: 1004703 |
| Element | Ammonia, dilute R3 |
| Quantity | 1000 mL |
| Grade | Eu. Ph. |