missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Amintra (R) Ni-NTA Affinity Resin

Product Code. p-7180224 Shop All Bio Techne Products
Click to view available options
Quantity:
10 mL
100 mL
Unit Size:
100mL
10mL
This item is not returnable. View return policy

Product Code. 18032446

Brand: Novus Biologicals™ ANN0010

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Highly purified and high bioactivity. Generating reliable and reproducible results.

  • Ni-NTA affinity resin is designed for simple, rapid His-tagged recombinant protein purification from a cell lysate under native or denaturing conditions. Metal chelate affinity chromatography is a rapid one-step purification, which removes most contaminants and can achieve purities close to homogeneity.
  • Offers excellent flow properties and high protein binding capacity: in excess of 50 mg protein/mL
  • NTA is a tetradentate chelator which occupies four of the six binding sites in the coordination sphere of the nickel ion. The other two coordination sites are usually occupied by water molecules and can interact with histidine residues of the recombinant protein. This binding minimizes metal leaching during purification.
  • This product is manufactured by Expedeon Inc. and distributed by Novus Biologicals.
  • This product is for research use only and is not approved for use in humans or in clinical diagnosis.
TRUSTED_SUSTAINABILITY

Specifications

Quantity 10 mL
Storage Requirements Store at 4°C. Do not freeze.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.