missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aminopeptidase N/CD13 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-77451SS
This item is not returnable.
View return policy
Description
Aminopeptidase N/CD13 Polyclonal antibody specifically detects Aminopeptidase N/CD13 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| Aminopeptidase N/CD13 | |
| Polyclonal | |
| Unconjugated | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 1.0 mg/mL | |
| Western Blot 0.5 - 2.0 μg/ml, Flow Cytometry 2-5 μg/ml, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| alanyl (membrane) aminopeptidase, Alanyl aminopeptidase, Aminopeptidase M, aminopeptidase N, AP-M, AP-N, CD13 antigen, CD13APN, EC 3.4.11, EC 3.4.11.2, gp150, LAP1, Microsomal aminopeptidase, Myeloid plasma membrane glycoprotein CD13, P150, PEPNhAPN | |
| Partial recombinant mouse CD13 protein (amino acids 69-966) [UniProt P97449] | |
| 0.025 mg | |
| Angiogenesis, Cellular Markers, Hematopoietic Stem Cell Markers, Immunology, Mesenchymal Stem Cell Markers, Myeloid derived Suppressor Cell, Stem Cell Markers | |
| 290 | |
| Store at 4°C for up to 3 months. For longer storage, aliquot and store at -20°C. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction