Learn More
Aminomethanesulfonic acid, 97%
Product Code. 11445417
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
Product Code. 11445417
Brand: Thermo Scientific Alfa Aesar B23441.14
Request Bulk or Custom Format
CAS: 13881-91-9 | CH5NO3S | 111.115 g/mol
Aminomethanesulfonic acid is used as a pharmaceutical intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Aminomethanesulfonic acid is used as a pharmaceutical intermediates.
Solubility
Soluble in water. (17 g/L) at 17°C
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are oxidizing agents. Store at room temperature.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 13881-91-9
Molecular Formula CH5NO3S
Molecular Weight (g/mol) 111.115
MDL Number MFCD00008124
InChI Key OBESRABRARNZJB-UHFFFAOYSA-N
Synonym methanesulfonic acid, amino, aminomethanesulphonic acid, aminomethyl sulfonic acid, aminomethylsulfonic acid, unii-ola224z482, methanesulfonic acid, 1-amino, aminomethanesulfonicacid, sodium aminomethanesulphonate, aminomethanesulfonic, aminomethanesulfoniccid
PubChem CID 83791
IUPAC Name aminomethanesulfonic acid
SMILES C(N)S(=O)(=O)O
Specifications
Melting Point ∼180°C (decomposition)
pH 3.2
Quantity 25 g
UN Number UN2585
Beilstein 1811756
Formula Weight 111.12
Percent Purity 97%
Chemical Name or Material Aminomethanesulfonic acid