accessibility menu, dialog, popup

UserName

CAS RN 13881-91-9

H2NSOHOO

CAS RN 13881-91-9

IUPAC Name: aminomethanesulfonic acid
Synonyms: aminomethanesulfonic acid
Molecular Weight (g/mol): 111.12
Molecular Formula: CH5NO3S
InChi Key: OBESRABRARNZJB-UHFFFAOYSA-N
SMILES: NCS(O)(=O)=O
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)