Learn More
Vitlab™ Amber Round Neck Reservoir Bottle
Click to view available options
Capacity (Metric):
2500 mL
Description
- Ethylene acrylate coating for increased safety
- Screw cap
- Plastic coating significantly reduces hazardous glass splintering during breakage
- 'Maximum working temperature for coated bottles is 80 °C.
- Preserve coating by not cleaning at temperatures exceeding 60 ºC.
Specifications
Capacity (Metric) | 2500 mL |
Coating | Plastic |
Color | Amber |
Material | Soda Lime Glass |
Max. Temperature (Metric) | 80°C |
Packaging | 1 Each |
Quantity | 1 Each |
Type | Bottle |
Closure Included | Yes |
Shape | Round |