missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AMACR/p504S Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15918-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes a racemase. The encoded enzyme interconverts pristanoyl-CoA and C27-bile acylCoAs between their (R)- and (S)-stereoisomers. The conversion to the (S)-stereoisomers is necessary for degradation of these substrates by peroxisomal beta-oxidation. Encoded proteins from this locus localize to both mitochondria and peroxisomes. Mutations in this gene may be associated with adult-onset sensorimotor neuropathy, pigmentary retinopathy, and adrenomyeloneuropathy due to defects in bile acid synthesis. Alternatively spliced transcript variants have been described.Specifications
| AMACR/p504S | |
| Polyclonal | |
| Unconjugated | |
| AMACR | |
| 2 methylacyl CoA racemase, alpha methylacyl CoA racemase, AMACR, CBAS4, p504S, RACE | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 23600 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q9UHK6 | |
| AMACR | |
| AMACR/p504S Fusion Protein Ag8720 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction