missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Alsin Polyclonal Antibody

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA5145109

Product Code. 18036211

  • 604.00€ / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: LGFWKTFPGK MTDSLRKPER RLLCESSNRA LSLQHAGRFS VNWFILFNDA LVHAQFSTHH VFPLATLWAE PLSEEAGGVN Antigen sequence identity: Mouse 98%; Rat 98%.

The function of this protein remains unknown.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Alsin
Polyclonal
Unconjugated
ALS2
3222402C23Rik; 9430073A21Rik; Als2; ALS2, alsin Rho guanine nucleotide exchange factor; Als2cr6; Alsin; ALSJ; amyotrophic lateral sclerosis 2 (juvenile); amyotrophic lateral sclerosis 2 (juvenile) homolog; amyotrophic lateral sclerosis 2 chromosomal region candidate gene 6 protein; Amyotrophic lateral sclerosis 2 protein; amyotrophic lateral sclerosis 2 protein homolog; amyotrophic lateral sclerosis protein 2 homolog; FLJ31851; IAHSP; KIAA1563; MGC87187; mKIAA1563; PLSJ
Rabbit
Affinity chromatography
RUO
57679
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
IgG
Immunocytochemistry
0.05 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q96Q42
ALS2
Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Antigen range: 893-972. Recombinant protein control fragment (Product #RP-109739).
100 μL
Primary
Human
Antibody
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.