Learn More
Ansell AlphaTec™ 1500 PLUS Lab Coats, Model 208
Product Code. p-7228392
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
Unit Size:
Pack of 70
Product Code. 17131005
Brand: Ansell WH15B0020805
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Short length lab coat with collar, elasticated cuffs, studded front fastening, left breast and right thigh pockets.
  • Air and water vapor permeable (breathable)
  • Silicone Free
  • Optimized body fit helps improve wearer comfort and safety
TRUSTED_SUSTAINABILITY
Specifications
Product Type General Purpose Lab Coat
Size X-Large
Gender Unisex
Material SMS
Color White
Closure Type Press Studs
Cuff Style Elastic
Disposable Reusable
Sterility Non-sterile
Resistance Chemicals
Style Short
No. of Pockets 1
Model W15B208
Seam Type Bound
Certifications/Compliance (Europe) EN ISO 13688:2013, Partial Body Protection Only, EN 13034:2005 + A1:2009
Weight (Metric) 48 gsm