missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AlphaB Crystallin/CRYAB Antibody (CL10758), Novus Biologicals™
Mouse Monoclonal Antibody
329.00€ - 516.00€
Specifications
| Antigen | AlphaB Crystallin/CRYAB |
|---|---|
| Clone | CL10758 |
| Dilution | Western Blot 1 ug/ml, Immunohistochemistry 1:20000 - 1:50000, Immunocytochemistry/Immunofluorescence 2-10 ug/ml, Immunohistochemistry-Paraffin 1:20000 - 1:50000 |
| Applications | Western Blot, Immunohistochemistry (Paraffin), Immunofluorescence, Immunohistochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18669644
|
Novus Biologicals
NBP3-21180-25ul |
25 ul |
329.00€
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18652664
|
Novus Biologicals
NBP3-21180-100ul |
100 μg |
516.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
AlphaB Crystallin/CRYAB Monoclonal antibody specifically detects AlphaB Crystallin/CRYAB in Human, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
| AlphaB Crystallin/CRYAB | |
| Western Blot 1 ug/ml, Immunohistochemistry 1:20000 - 1:50000, Immunocytochemistry/Immunofluorescence 2-10 ug/ml, Immunohistochemistry-Paraffin 1:20000 - 1:50000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Rat, Human | |
| alpha B crystallin, alpha(B)-crystallin, alpha-crystallin B chain, CRYA2alpha crystallin B chain, crystallin, alpha B, CTPP2, Heat shock protein beta-5, heat-shock 20 kD like-protein, HSPB5, Renal carcinoma antigen NY-REN-27, Rosenthal fiber component | |
| This antibody was developed against a Synthetic Peptide of P02511. Actual immunogen sequence is proprietary. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| CL10758 | |
| Western Blot, Immunohistochemistry (Paraffin), Immunofluorescence, Immunohistochemistry | |
| Unconjugated | |
| Mouse | |
| Cancer, Cell Biology, Cell Cycle and Replication, Golgi Apparatus Markers, Membrane Trafficking and Chaperones, Neuroscience, Neurotransmission, Phospho Specific, Protein Phosphatase, Sensory Systems, Signal Transduction, Tyrosine Kinases, Vision | |
| PBS, pH 7.2, 40% glycerol | |
| 1410 | |
| IgG1 | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title