missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AlphaA Crystallin/CRYAA Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-05575-20ul
This item is not returnable.
View return policy
Description
AlphaA Crystallin/CRYAA Polyclonal antibody specifically detects AlphaA Crystallin/CRYAA in Human, Mouse, Rat, Zebrafish samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications
| AlphaA Crystallin/CRYAA | |
| Polyclonal | |
| Western Blot 1:2000 - 1:6000, Immunocytochemistry/ Immunofluorescence 1:50 - 1:200 | |
| CRYA1crystallin, alpha-1, crystallin, alpha A, Heat shock protein beta-4, HSPB4alpha-crystallin A chain, human alphaA-crystallin (CRYA1), 10HspB4 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-173 of human AlphaA Crystallin/CRYAA (NP_000385.1). MDVTIQHPWFKRTLGPFYPSRLFDQFFGEGLFEYDLLPFLSSTISPYYRQSLFRTVLDSGISEVRSDRDKFVIFLDVKHFSPEDLTVKVQDDFVEIHGKHNERQDDHGYISREFHRRYRLPSNVDQSALSCSLSADGMLTFCGPKIQTGLDATHAERAIPVSREEKPTSAPSS | |
| 20 μg | |
| Membrane Trafficking and Chaperones, Vision | |
| 1409 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Zebrafish | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction