missing translation for 'onlineSavingsMsg'
Learn More
Learn More
alpha Tubulin Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
339.00€ - 548.00€
Specifications
| Antigen | alpha Tubulin |
|---|---|
| Dilution | Western Blot 0.04-0.4 μg/mL, Immunohistochemistry 1:500 - 1:1000, Immunocytochemistry/Immunofluorescence 0.25-2 μg/mL, Immunohistochemistry-Paraffin 1:500 - 1:1000 |
| Applications | Western Blot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18287290
|
Novus Biologicals
NBP2-54687 |
100 μL |
548.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18613189
|
Novus Biologicals
NBP2-54687-25ul |
25 μL |
339.00€
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
alpha Tubulin Polyclonal specifically detects alpha Tubulin in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| alpha Tubulin | |
| Western Blot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human, Mouse, Rat | |
| Alpha-tubulin 3, B-ALPHA-1, FLJ25113, hum-a-tub1, hum-a-tub2, LIS3, TUBA3, tubulin alpha-1A chain, Tubulin alpha-3 chain, Tubulin B-alpha-1, tubulin, alpha 1a, tubulin, alpha 3, tubulin, alpha, brain-specific | |
| TUBA1A | |
| IgG | |
| Affinity Purified | |
| 50 kDa |
| Western Blot 0.04-0.4 μg/mL, Immunohistochemistry 1:500 - 1:1000, Immunocytochemistry/Immunofluorescence 0.25-2 μg/mL, Immunohistochemistry-Paraffin 1:500 - 1:1000 | |
| Polyclonal | |
| Rabbit | |
| Cell Biology, Cell Cycle and Replication, Cytoskeleton Markers, Loading Controls, Neuroscience | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
| 7846 | |
| This alpha Tubulin Antibody was developed against a recombinant protein corresponding to amino acids: LEHGIQPDGQMPSDKTIGGGDDSFNTFF | |
| Primary | |
| The specificity of this human alpha Tubulin Antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title