missing translation for 'onlineSavingsMsg'
Learn More
Learn More
alpha-SNAP Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10546-1-AP-20UL
This item is not returnable.
View return policy
Description
This antibody recognizes a 33 kDa protein in SDS page.
SNAPs (solubleNSFattachment proteins), acting in concert with SNAREs (S N A P r e c eptors) and the N - ethylmaleimide- s e nsitive f u sion protein (NSF) are required for the fusion of transport vesicles to their target membranes in synaptic transmission, intra-Golgi transport, endosome-toendosome fusion and transcytotic vesicles-to-plasma membrane transport. Vesicle-to-target membrane docking (initial contact) occurs when the vesicle SNARE binds to its cognate target membrane SNARE. a-SNAP (or b-SNAP in brain) then binds to this docking complex and mediates the binding of NSF and thus the formation of a 20S fusion particle. It is thought that, once NSF is bound, ATP hydrolysis by NSF initiates the fusion process.Specifications
| alpha-SNAP | |
| Polyclonal | |
| Unconjugated | |
| NAPA | |
| alpha-SNAP, NAPA, SNAP alpha, SNAPA | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 108124, 140673, 8775 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.23 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P54920, P54921, Q9DB05 | |
| NAPA | |
| alpha-SNAP Fusion Protein Ag0836 | |
| 20 μL | |
| Primary | |
| Rat, Mouse, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction