Learn More
alpha-Phenylcyclopentylacetic acid, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
Description
alpha-Phenylcyclopentylacetic acid is used as pharmaceutical intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
alpha-Phenylcyclopentylacetic acid is used as pharmaceutical intermediates.
Solubility
Insoluble in water.
Notes
Store in a cool and dark place. Store away from incompatible materials such as oxidizing agents.
alpha-Phenylcyclopentylacetic acid is used as pharmaceutical intermediates.
Solubility
Insoluble in water.
Notes
Store in a cool and dark place. Store away from incompatible materials such as oxidizing agents.
Specifications
Melting Point | 98°C to 102°C |
Quantity | 25 g |
Beilstein | 1956835 |
Solubility Information | Insoluble in water. |
Formula Weight | 204.27 |
Percent Purity | 98% |
Chemical Name or Material | alpha-Phenylcyclopentylacetic acid |