missing translation for 'onlineSavingsMsg'
Learn More

alpha-Hydroxyalprazolam, Mikromol™

Product Code. 17980436
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17980436 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17980436 Supplier Mikromol Supplier No. MM0136.02

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

alpha-Hydroxyalprazolam
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material alpha-Hydroxyalprazolam
CAS 37115-43-8
Analyte or Component Names alpha-Hydroxyalprazolam
Shipping Condition Room Temperature
Molecular Formula C17 H13 Cl N4 O
Certifications/Compliance ISO 17025
Synonym alpha-Hydroxyalprazolam, -[1,2,4]Triazolo[4,3-a][1,4]benzodiazepine-1-methanol, 8-chloro-6-phenyl-, 1-Hydroxyalprazolam, 1'-Hydroxyalprazolam, 6-Phenyl-8-chloro-4H-s-triazolo[4,3-a]benzodiazepine-1-methanol, U 40125
InChI Formula InChI=1S/C17H13ClN4O/c18-12-6-7-14-13(8-12)17(11-4-2-1-3-5-11)19-9-15-20-21-16(10-23)22(14)15/h1-8,23H,9-10H2
SMILES OCc1nnc2CN=C(c3ccccc3)c4cc(Cl)ccc4n12
IUPAC Name (8-chloro-6-phenyl-4H-[1,2,4]triazolo[4,3-a][1,4]benzodiazepin-1-yl)methanol
Molecular Weight (g/mol) 324.76
Quantity 25 mg
Formula Weight 324.0778 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.