missing translation for 'onlineSavingsMsg'
Learn More

alpha-Angelicalactone, 98%

Product Code. 11331977
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11331977

Brand: Thermo Scientific Alfa Aesar L07602.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 591-12-8 | C5H6O2 | 98.10 g/mol

alpha-Angelicalactone acts as acancer chemopreventive agent. It is used in the synthesis of glutathione and the activity of glutathione-S-transferase and UDP-glucunonosyltransferase detoxification enzymes in esophagus, stomach, intestine, and liver. It is used in oral care formulations to round out mint notes and in coffee flavors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
alpha-Angelicalactone acts as acancer chemopreventive agent. It is used in the synthesis of glutathione and the activity of glutathione-S-transferase and UDP-glucunonosyltransferase detoxification enzymes in esophagus, stomach, intestine, and liver. It is used in oral care formulations to round out mint notes and in coffee flavors.

Solubility
Soluble in water (5g/100mL) and alcohol.

Notes
Moisture-sensitive. Store under inert gas. Protect from moisture. Incompatible materials are Oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 591-12-8
Molecular Formula C5H6O2
Molecular Weight (g/mol) 98.10
MDL Number MFCD00005375
InChI Key QOTQFLOTGBBMEX-UHFFFAOYSA-N
Synonym alpha-angelica lactone, alpha-angelicalactone, 5-methyl-2 3h-furanone, 5-methylfuran-2 3h-one, 2 3h-furanone, 5-methyl, 4-hydroxypent-3-enoic acid lactone, delta 2-angelica lactone, 4-hydroxy-3-pentenoic acid gamma-lactone, gamma-methyl-beta,gamma-crotonolactone, .alpha.-angelica lactone
PubChem CID 11559
ChEBI CHEBI:36433
IUPAC Name 5-methyl-3H-furan-2-one
SMILES CC1=CCC(=O)O1

Specifications

Melting Point 17°C to 18°C
Density 1.093
Boiling Point 53°C to 54°C (10 mmHg)
Flash Point 68°C (154°F)
Refractive Index 1.448
Quantity 25 g
Beilstein 108394
Merck Index 14,647
Solubility Information Soluble in water (5g/100mL) and alcohol.
Formula Weight 98.1
Percent Purity 98%
Chemical Name or Material alpha-Angelicalactone
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.