missing translation for 'onlineSavingsMsg'
Learn More
Learn More
alpha Adducin Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
228.00€ - 315.00€
Specifications
| Antigen | alpha Adducin |
|---|---|
| Dilution | Immunohistochemistry-Paraffin 1:200-1:1000 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18636576
|
Novus Biologicals
NBP2-99388-100ul |
100 μg |
315.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18637797
|
Novus Biologicals
NBP2-99388-50ul |
50 μg |
228.00€
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
alpha Adducin Polyclonal antibody specifically detects alpha Adducin in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
| alpha Adducin | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Human | |
| ADDA, adducin 1 (alpha), alpha-adducin, erythrocyte adducin alpha subunit, Erythrocyte adducin subunit alpha, MGC3339, MGC44427 | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human alpha Adducin. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Immunohistochemistry-Paraffin 1:200-1:1000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.0) | |
| 118 | |
| IgG | |
| Antigen and protein A Affinity-purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title