missing translation for 'onlineSavingsMsg'
Learn More

alpha-1B-glycoprotein Mouse anti-Human, Clone: 4D4E5, Proteintech

Mouse Monoclonal Antibody

Brand:  Proteintech 66260-1-IG-150UL

 View more versions of this product

Product Code. 16817975

  • 410.00€ / 150µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Alpha-1-B glycoprotein (A1BG) which is a plasma protein with no known function belongs to the immunoglobulin superfamily. Although the human A1BG has been known for four decades, and the information about the amino acid sequence, chromosomal assignment, and even genetic polymorphism in different populations have been known, no biological function has been suggested. A1BG (Mr approximately equal to 63,000) consists of one polypeptide chain of 474 amino acids with four glucosamine oligosaccharides. The polypeptide has five intrachain disulfide bonds and consists of five repeating structural domains, each containing about 95 amino acids and one disulfide bond. Recently human cyctein-rich secretory protein 3 (CRISP-3) has been found to be a binding partner of A1B, suggesting the A1BG-CRISP-3 complex displays a protecting function from a potentially harmful effect of free CRISP-3 in circulation.
Specifications

Specifications

alpha-1B-glycoprotein
Monoclonal
0.89 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P04217
A1BG
alpha-1B-glycoprotein Fusion Protein Ag5414
150 μL
Primary
Human
Antibody
IgG1
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
4D4E5
Unconjugated
A1BG
A1B, A1BG, ABG, alpha 1 B glycoprotein, Alpha 1B glycoprotein, GAB, HYST2477
Mouse
Protein A
RUO
1
-20°C
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.