missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Allophycocyanin Antibody (936809) [Allophycocyanin], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB8927A
This item is not returnable.
View return policy
Description
Allophycocyanin Monoclonal antibody specifically detects Allophycocyanin in Broad Species Reactivity samples. It is validated for Flow Cytometry, CyTOF
Specifications
| Allophycocyanin | |
| Monoclonal | |
| APC | |
| PBS | |
| Mouse | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| Broad Species Reactivity | |
| Purified |
| Flow Cytometry, CyTOF | |
| 936809 | |
| Flow Cytometry, CyTOF-ready | |
| APC | |
| Allophycocyanin isolated from red algae, Accession # P25054 | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction