missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Allergin 1 Monoclonal Antibody (767727)
GREENER_CHOICE

Mouse Monoclonal Antibody

Brand:  Invitrogen™ MA561849

Product Code. 30535297

  • 439.00€ / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Reconstitute with sterile PBS to a final concentration of 0.5 mg/mL.

Allergin 1 is a protein coding gene. Immunoglobulin-like receptor which plays an inhibitory role in degranulation of mast cells. Negatively regulates IgE-mediated mast cell activation and suppresses the type I immediate hypersensitivity reaction.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Allergin 1
Monoclonal
0.5 mg/mL
PBS with trehalose and no preservative
Q7Z6M3
MILR1
Mouse myeloma cell line NS0-derived recombinant human Allergin-1, Met1-Lys227.
100 μg
Primary
Human
Antibody
IgG1
Flow Cytometry, Immunocytochemistry
767727
Unconjugated
MILR1
Allergin-1; Allergy inhibitory receptor 1; C17orf60; Gm885; mast cell Ag-32; mast cell antigen 32; mast cell immunoglobulin like receptor 1; mast cell immunoglobulin-like receptor 1; MCA32; MCA-32; MCA32-1S2; Milr1; probable mast cell antigen 32 homolog; surface protein MCA-32
Mouse
Protein A/G
RUO
284021
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Lyophilized
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.