missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ALKBH7 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
190.00€ - 463.00€
Specifications
| Antigen | ALKBH7 |
|---|---|
| Dilution | Western Blot 1:200-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18651621
|
Novus Biologicals
NBP2-92409-0.02ml |
0.02 mL |
190.00€
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18640731
|
Novus Biologicals
NBP2-92409-0.1ml |
0.1 mL |
463.00€
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ALKBH7 Polyclonal antibody specifically detects ALKBH7 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| ALKBH7 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Direct Reversal of DNA Damage | |
| PBS with 50% glycerol, pH7.3. | |
| 84266 | |
| IgG | |
| Affinity purified |
| Western Blot 1:200-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| ABH7SPATA11, alkB, alkylation repair homolog 7 (E. coli), Alkylated DNA repair protein alkB homolog 7, EC 1.14.11.-, MGC10974, probable alpha-ketoglutarate-dependent dioxygenase ABH7, spermatogenesis associated 11, spermatogenesis cell proliferation related protein, Spermatogenesis cell proliferation-related protein, Spermatogenesis-associated protein 11, UNQ6002 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-221 of human ALKBH7 (NP_115682.1). MAGTGLLALRTLPGPSWVRGSGPSVLSRLQDAAVVRPGFLSTAEEETLSRELEPELRRRRYEYDHWDAAIHGFRETEKSRWSEASRAILQRVQAAAFGPGQTLLSSVHVLDLEARGYIKPHVDSIKFCGATIAGLSLLSPSVMRLVHTQEPGEWLELLLEPGSLYILRGSARYDFSHEILRDEESFFGERRIPRGRRISVICRSLPEGMGPGESGQPPPAC | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title