missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alkaline Phosphatase/ALPP Antibody (SPM593), Novus Biologicals™
Mouse Monoclonal Antibody
290.00€ - 511.00€
Specifications
| Antigen | Alkaline Phosphatase/ALPP |
|---|---|
| Clone | SPM593 |
| Concentration | 0.2mg/mL |
| Dilution | Flow Cytometry 0.5 - 1 ug/million cells in 0.1 ml, Immunohistochemistry-Paraffin 0.25 - 0.5 ug/ml, Immunofluorescence 0.5 - 1.0 ug/ml |
| Applications | Flow Cytometry, Immunohistochemistry (Paraffin), Immunofluorescence |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18170809
|
Novus Biologicals
NBP2-44955-0.02MG |
0.02 mg |
290.00€
0.02mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18074524
|
Novus Biologicals
NBP2-44955-0.1MG |
0.1 mg |
511.00€
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Ensure accurate, reproducible results in Flow Cytometry, Immunohistochemistry (Paraffin), Immunofluorescence
Alkaline Phosphatase/ALPP Monoclonal specifically detects Alkaline Phosphatase/ALPP in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunofluorescence.Specifications
| Alkaline Phosphatase/ALPP | |
| 0.2mg/mL | |
| Flow Cytometry, Immunohistochemistry (Paraffin), Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Cancer, Embryonic Stem Cell Markers, Ovarian Carcinoma Cell Markers, Stem Cell Markers | |
| 10mM PBS and 0.05% BSA with 0.05% Sodium Azide | |
| Alkaline phosphatase Regan isozyme, alkaline phosphatase, placental, alkaline phosphatase, placental (Regan isozyme), alkaline phosphatase, placental type, alkaline phosphomonoesterase, ALP, EC 3.1.3.1, FLJ61142, glycerophosphatase, PALP, Placental alkaline phosphatase 1, PLAP, PLAP-1, Regan isozyme | |
| ALPP | |
| IgG2b κ | |
| Protein A or G purified | |
| Reacts with a 70kDa membrane-bound isozyme (Regan and Nagao type) of Placental Alkaline Phosphatase (PLAP) occurring in the placenta during the 3rd trimester of gestation. It is highly specific for PLAP and shows no cross-reaction with other isozymes of alkaline phosphatase. Anti-PLAP reacts with germ cell tumors and can discriminate between these and other neoplasms. Somatic neoplasms e.g. breast, gastrointestinal, prostatic, and urinary cancers may also immunoreact with antibodies to PLAP. Anti-PLAP positivity in conjunction with anti-keratin negativity favors seminoma over carcinoma. Germ cell tumors are usually anti-keratin positive, but they regularly fail to stain with anti-EMA, whereas most carcinomas stain with anti-EMA. Anti-PLAP has been useful in the diagnosis of gestational trophoblastic disease |
| SPM593 | |
| Flow Cytometry 0.5 - 1 ug/million cells in 0.1 ml, Immunohistochemistry-Paraffin 0.25 - 0.5 ug/ml, Immunofluorescence 0.5 - 1.0 ug/ml | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| P05187 | |
| 250 | |
| Recombinant full-length human ALPP protein | |
| Primary | |
| Store at 4C. | |
| 70 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title