missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Alkaline Phosphatase/ALPP Antibody (Seagen Patent Anti-Alpp) - Humanized, Novus Biologicals™
Human Monoclonal Antibody
213.00€ - 920.00€
Specifications
| Antigen | Alkaline Phosphatase/ALPP |
|---|---|
| Clone | Seagen Patent Anti-Alpp |
| Applications | Flow Cytometry, ELISA, Functional Assay |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30046234
|
Novus Biologicals™
NBP3-28189-50ug |
50 μg |
213.00€
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30047034
|
Novus Biologicals™
NBP3-28189-1mg |
1 mg |
920.00€
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30046590
|
Novus Biologicals™
NBP3-28189-100ug |
100 μg |
338.00€
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Alkaline Phosphatase/ALPP Monoclonal antibody specifically detects Alkaline Phosphatase/ALPP in Human samples. It is validated for Flow Cytometry,ELISA,Functional AssaySpecifications
| Alkaline Phosphatase/ALPP | |
| Flow Cytometry, ELISA, Functional Assay | |
| Unconjugated | |
| Human | |
| Cancer, Ovarian Carcinoma Cell Markers, Phospho Specific, Stem Cell Markers | |
| Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) | |
| 250 | |
| IgG1 | |
| Protein A purified | |
| Store at -20°C in powder form. Store at -80°C once reconstituted. |
| Seagen Patent Anti-Alpp | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| Alkaline phosphatase Regan isozyme, alkaline phosphatase, placental, alkaline phosphatase, placental (Regan isozyme), alkaline phosphatase, placental type, alkaline phosphomonoesterase, ALP, EC 3.1.3.1, FLJ61142, glycerophosphatase, PALP, Placental alkaline phosphatase 1, PLAP, PLAP-1, Regan isozyme | |
| ALP | |
| Primary | |
| Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title