missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Alkaline Phosphatase/ALPP Monoclonal specifically detects Alkaline Phosphatase/ALPP in Human, Mouse, Rat samples. It is validated for Flow Cytometry.
Specifications
Specifications
| Antigen | Alkaline Phosphatase/ALPP |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 8B6 |
| Conjugate | PE |
| Dilution | Flow Cytometry |
| Gene Alias | Alkaline phosphatase Regan isozyme, alkaline phosphatase, placental, alkaline phosphatase, placental (Regan isozyme), alkaline phosphatase, placental type, alkaline phosphomonoesterase, ALP, EC 3.1.3.1, FLJ61142, glycerophosphatase, PALP, Placental alkaline phosphatase 1, PLAP, PLAP-1, Regan isozyme |
| Gene Symbols | ALPP |
| Host Species | Mouse |
| Immunogen | Hep-2 cells with boosted surface expression of Alkaline Phosphatase, Placental. [UniProt# P05187]. |
| Show More |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?