missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aldolase C Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14884-1-AP-20UL
This item is not returnable.
View return policy
Description
This gene encodes a member of the class I fructose-biphosphate aldolase gene family. Expressed specifically in the hippocampus and Purkinje cells of the brain, the encoded protein is a glycolytic enzyme that catalyzes the reversible aldol cleavage of fructose-1, 6-biphosphate and fructose 1-phosphate to dihydroxyacetone phosphate and either glyceraldehyde-3-phosphate or glyceraldehyde, respectively.Specifications
| Aldolase C | |
| Polyclonal | |
| Unconjugated | |
| ALDOC | |
| ALDOC, Aldolase C, Brain type aldolase, Fructose bisphosphate aldolase C | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 11676, 230, 24191 | |
| -20°C | |
| Liquid |
| Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry, Immunofluorescence | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P05063, P09117, P09972 | |
| ALDOC | |
| Aldolase C Fusion Protein Ag6659 | |
| 20 μL | |
| Primary | |
| Rat, Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction