Learn More
E2F-4 Antibody (24E12), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals NBP3-26608-100ul
 View more versions of this product
Product Code. 30043921
  • 533.00€ / 100µL
Description
E2F-4 Monoclonal antibody specifically detects E2F-4 in Human samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry
TRUSTED_SUSTAINABILITY
Specifications
Specifications
E2F-4
Monoclonal
Unconjugated
PBS, pH 7.4, 150mM NaCl and 50% glycerol
Rabbit
Affinity purified
RUO
Primary
Human
Purified
Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry
24E12
Flow Cytometry 1:50-1:200, ELISA, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200
E2F transcription factor 4, p107/p130-binding, E2F-4p107/p130-binding protein, transcription factor E2F4
A synthesized peptide derived from Human E2F-4 [UniProt Q16254]
100 μL
Cell Cycle and Replication
1874
Store at -20 to -70C. Avoid freeze-thaw cycles.