missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ALDOA Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11217-1-AP-20UL
This item is not returnable.
View return policy
Description
Aldolase A (fructose-bisphosphate aldolase) is a glycolytic enzyme that catalyzes the reversible conversion of fructose-1, 6-bisphosphate to glyceraldehyde 3-phosphate and dihydroxyacetone phosphate. Three aldolase isozymes (A, B, and C), encoded by three different genes, are differentially expressed during development. Aldolase A is found in the developing embryo and is produced in even greater amounts in adult muscle. Aldolase A expression is repressed in adult liver, kidney and intestine and similar to aldolase C levels in brain and other nervous tissue. Aldolase A deficiency has been associated with myopathy and hemolytic anemia.Specifications
| ALDOA | |
| Polyclonal | |
| Unconjugated | |
| ALDOA | |
| ALDOA, Aldolase A | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 11674, 226, 24189 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry | |
| 0.23 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P04075, P05064, P05065 | |
| ALDOA | |
| ALDOA Fusion Protein Ag1726 | |
| 20 μL | |
| Primary | |
| Rat, Mouse, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction