missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Aldo-keto Reductase 1C1/AKR1C1 Antibody (AKR1C1/9070), Novus Biologicals™
Mouse Monoclonal Antibody
232.00€ - 462.00€
Specifications
| Antigen | Aldo-keto Reductase 1C1/AKR1C1 |
|---|---|
| Clone | AKR1C1/9070 |
| Concentration | 0.2 mg/mL |
| Dilution | Immunohistochemistry-Paraffin 1-2 μg/mL for 30 minutes at RT |
| Applications | Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30043275
|
Novus Biologicals™
NBP3-26772-100ug |
100 μg |
462.00€
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30043671
|
Novus Biologicals™
NBP3-26772-20ug |
20 μg |
232.00€
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Aldo-keto Reductase 1C1/AKR1C1 Monoclonal antibody specifically detects Aldo-keto Reductase 1C1/AKR1C1 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
| Aldo-keto Reductase 1C1/AKR1C1 | |
| 0.2 mg/mL | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| metabolism | |
| 10 mM PBS with 0.05% BSA | |
| 1645 | |
| IgG2b κ | |
| Protein A or G purified |
| AKR1C1/9070 | |
| Immunohistochemistry-Paraffin 1-2 μg/mL for 30 minutes at RT | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| 20 alpha-hydroxysteroid dehydrogenase, 20-ALPHA-HSD, 20-alpha-hydroxysteroid dehydrogenase, aldo-keto reductase family 1 member C1, aldo-keto reductase family 1, member C1 (dihydrodiol dehydrogenase 1; 20-alpha(3-alpha)-hydroxysteroid dehydrogenase), C9, Chlordecone reductase homolog HAKRC, DD1/DD2, DD1MGC8954, DDHH-37, dihydrodiol dehydrogenase 1, Dihydrodiol dehydrogenase 1/2, dihydrodiol dehydrogenase isoform DD1, EC 1.1.1, EC 1.1.1.-, EC 1.1.1.112, EC 1.1.1.149,2-ALPHA-HSD, EC 1.3.1.20, HAKRCDDH1aldo-keto reductase C, HBAB, hepatic dihydrodiol dehydrogenase, High-affinity hepatic bile acid-binding protein, Indanol dehydrogenase, MBAB, Trans-1,2-dihydrobenzene-1,2-diol dehydrogenase, type II 3-alpha-hydroxysteroid dehydrogenase | |
| Recombinant human full-length Aldo-keto Reductase 1C1/AKR1C1 protein | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title