missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ALDH7A1 Rabbit anti-Human, Clone: 105, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89927-100UL
This item is not returnable.
View return policy
Description
ALDH7A1 Monoclonal specifically detects ALDH7A1 in Human samples. It is validated for Western Blot, ELISA, Immunoprecipitation.
Specifications
| ALDH7A1 | |
| Monoclonal | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA, Immunoprecipitation | |
| 105 | |
| Western Blot 1:1000-1:5000, ELISA 1:5000-1:10000, Immunoprecipitation 1-4 ul/mg of lysate | |
| aldehyde dehydrogenase 7 family, member A1, Aldehyde dehydrogenase family 7 member A1,26g turgor protein homolog, Alpha-AASA dehydrogenase, ATQ1Antiquitin-1, Betaine aldehyde dehydrogenase, Delta1-piperideine-6-carboxylate dehydrogenase, EC 1.2.1, EC 1.2.1.3, EC 1.2.1.31, EC 1.2.1.8, EPDalpha-aminoadipic semialdehyde dehydrogenase, FLJ11738, FLJ92814, P6c dehydrogenase, PDEantiquitin-1 | |
| Recombinant Human ALDH7A1 protein | |
| 100 μg | |
| Core ESC Like Genes, Stem Cell Markers | |
| 501 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction