missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ALDH1A3 Antibody (CL11968) [Janelia Fluor™ 525], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-15126JF525
This item is not returnable.
View return policy
Description
ALDH1A3 Monoclonal antibody specifically detects ALDH1A3 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
Specifications
| ALDH1A3 | |
| Monoclonal | |
| Janelia Fluor 525 | |
| aldehyde dehydrogenase 1 family, member A3, Aldehyde dehydrogenase 6, aldehyde dehydrogenase family 1 member A3, ALDH1A6, ALDH6acetaldehyde dehydrogenase 6, EC 1.2.1, EC 1.2.1.5, RalDH3, RALDH-3, Retinaldehyde dehydrogenase 3 | |
| This antibody was developed using a synthetic peptide derived from P47895, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Vision | |
| 220 | |
| Store at 4⪚C in the dark. | |
| IgG2a |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
| CL11968 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction