missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ALDH1A3 Antibody (CL11968) [DyLight 405], Novus Biologicals™
Mouse Monoclonal Antibody
Marca: Novus Biologicals NBP3-15126V
Les retours ne sont pas autorisés pour ce produit.
Consulta la politica di reso
Descrizione
ALDH1A3 Monoclonal antibody specifically detects ALDH1A3 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
Specifica
| ALDH1A3 | |
| Monoclonal | |
| DyLight 405 | |
| aldehyde dehydrogenase 1 family, member A3, Aldehyde dehydrogenase 6, aldehyde dehydrogenase family 1 member A3, ALDH1A6, ALDH6acetaldehyde dehydrogenase 6, EC 1.2.1, EC 1.2.1.5, RalDH3, RALDH-3, Retinaldehyde dehydrogenase 3 | |
| This antibody was developed using a synthetic peptide derived from P47895, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Vision | |
| 220 | |
| Store at 4⪚C in the dark. | |
| IgG2a |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
| CL11968 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto