missing translation for 'onlineSavingsMsg'
Learn More

Aldehyde dehydrogenase 5 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody

190.00€ - 463.00€

Specifications

Antigen Aldehyde dehydrogenase 5
Dilution Western Blot 1:500-1:2000, Immunocytochemistry/ Immunofluorescence 1:50-1:200
Applications Western Blot, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18640412
View Documents
Novus Biologicals
NBP2-92625-0.02ml
0.02 mL
190.00€
0.02mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18655052
View Documents
Novus Biologicals
NBP2-92625-0.1ml
0.1 mL
463.00€
0.10mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Aldehyde dehydrogenase 5 Polyclonal antibody specifically detects Aldehyde dehydrogenase 5 in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications

Specifications

Aldehyde dehydrogenase 5
Western Blot, Immunofluorescence
Unconjugated
Rabbit
Cancer, Endocrinology, Signal Transduction
PBS with 50% glycerol, pH7.3.
219
IgG
Affinity purified
Western Blot 1:500-1:2000, Immunocytochemistry/ Immunofluorescence 1:50-1:200
Polyclonal
Purified
RUO
Human
aldehyde dehydrogenase 1 family, member B1, Aldehyde dehydrogenase 5, Aldehyde dehydrogenase family 1 member B1, ALDH class 2, ALDH5aldehyde dehydrogenase X, mitochondrial, ALDHXacetaldehyde dehydrogenase 5, EC 1.2.1, EC 1.2.1.3, MGC2230, mitochondrial aldehyde dehydrogenase X
Recombinant fusion protein containing a sequence corresponding to amino acids 278-517 of human ALDH1B1 (NP_000683.3). NLKRVTLELGGKSPSIVLADADMEHAVEQCHEALFFNMGQCCCAGSRTFVEESIYNEFLERTVEKAKQRKVGNPFELDTQQGPQVDKEQFERVLGYIQLGQKEGAKLLCGGERFGERGFFIKPTVFGGVQDDMRIAKEEIFGPVQPLFKFKKIEEVVERANNTRYGLAAAVFTRDLDKAMYFTQALQAGTVWVNTYNIVTCHTPFGGFKESGNGRELGEDGLKAYTEVKTVTIKVPQKNS
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.