missing translation for 'onlineSavingsMsg'
Learn More
Learn More
alcohol dehydrogenase 5 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35343-20ul
This item is not returnable.
View return policy
Description
alcohol dehydrogenase 5 Polyclonal antibody specifically detects alcohol dehydrogenase 5 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot,Immunocytochemistry/Immunofluorescence
Specifications
| alcohol dehydrogenase 5 | |
| Polyclonal | |
| Western Blot 1:100 - 1:500, ELISA, Immunocytochemistry/Immunofluorescence 1:50 - 1:200 | |
| ADH-3, ADHXEC 1.1.1.1, alcohol dehydrogenase (class III), chi polypeptide, Alcohol dehydrogenase 5, alcohol dehydrogenase 5 (class III), chi polypeptide, Alcohol dehydrogenase class chi chain, alcohol dehydrogenase class-3, Alcohol dehydrogenase class-III, EC 1.1.1, EC 1.1.1.-, EC 1.1.1.284, FALDH, FDHGSNOR, formaldehyde dehydrogenase, Glutathione-dependent formaldehyde dehydrogenase, GSH-FDH, S-(hydroxymethyl)glutathione dehydrogenase | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-374 of human alcohol dehydrogenase 5 (NP_000662.3).,, Sequence:, MANEVIKCKAAVAWEAGKPLSIEEIEVAPPKAHEVRIKIIATAVCHTDAYTLSGADPEGCFPVILGHEGAGIVESVGEGVTKLKAGDTVIPLYIPQCGECKFCLNPKTNLCQKIRVTQGKGLMPDGTSRFTCKGKTILHYMGTSTFSEYTVVADISVAKIDPLAPLDKVCLLGCGISTGYGAAVNTAKLEPGSVCAVFGLGGVGLAVIMGCKVAGASRIIGVDINKDKFARAKEFGATECINPQDFSKPIQEVLIEMTDGGVDYSFECIGNVKVMRAALEACHKGWGVSVVVGVAASGEEIATRPFQLVTGRTWKGTAFGGWKSVESVPKLVSEYMSKKIKVDEFVTHNLSFDEINKAFELMHSGKSIRTVVKI | |
| 20 μL | |
| Core ESC Like Genes, Stem Cell Markers | |
| 128 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot, Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction