missing translation for 'onlineSavingsMsg'
Learn More
Learn More
alcohol dehydrogenase 4 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-38155-20ul
This item is not returnable.
View return policy
Description
alcohol dehydrogenase 4 Polyclonal antibody specifically detects alcohol dehydrogenase 4 in Mouse samples. It is validated for ELISA,Western Blot
Specifications
| alcohol dehydrogenase 4 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA | |
| ADH-2, alcohol dehydrogenase 4, alcohol dehydrogenase 4 (class II), pi polypeptide, Alcohol dehydrogenase class II pi chain, aldehyde reductase, EC 1.1.1, EC 1.1.1.1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-380 of human alcohol dehydrogenase 4 (NP_000661.2).,, Sequence:, MGTKGKVIKCKAAIAWEAGKPLCIEEVEVAPPKAHEVRIQIIATSLCHTDATVIDSKFEGLAFPVIVGHEAAGIVESIGPGVTNVKPGDKVIPLYAPLCRKCKFCLSPLTNLCGKISNLKSPASDQQLMEDKTSRFTCKGKPVYHFFGTSTFSQYTVVSDINLAKIDDDANLERVCLLGCGFSTGYGAAINNAKVTPGSTCAVFGLGGVGLSAVMGCKAAGASRIIGIDINSEKFVKAKALGATDCLNPRDLHKPIQEVIIELTKGGVDFALDCAGGSETMKAALDCTTAGWGSCTFIGVAAGSKGLTIFPEELIIGRTINGTFFGGWKSVDSIPKLVTDYKNKKFNLDALVTHTLPFDKISEAFDLMNQGKSVRTILIF | |
| 20 μL | |
| Lipid and Metabolism | |
| 127 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction