missing translation for 'onlineSavingsMsg'
Learn More

Alclofenac, TRC

Product Code. 30135891
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30135891 10 mg 10mg
30132946 100 mg 100mg
17974395 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30135891 Supplier Toronto Research Chemicals Supplier No. TRCA51455010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22131-79-9
Molecular Formula C11 H11 Cl O3
Molecular Weight (g/mol) 226.66
Synonym Benzeneacetic acid, 3-chloro-4-(2-propen-1-yloxy)-Acetic acid, [4-(allyloxy)-3-chlorophenyl]- (8CI), Benzeneacetic acid, 3-chloro-4-(2-propenyloxy)- (9CI), Aclofenac, Alclofenac, Alclophenac, Allopydin, Epinal, MY 101, Medifenac, Mervan, Neosten, Neoston, Prinalgin, Reufenac, W 7320, Zumaril, [3-Chloro-4-(allyloxy)phenyl]acetic acid, [4-(Allyloxy)-3-chlorophenyl]acetic acid
IUPAC Name 2-(3-chloro-4-prop-2-enoxyphenyl)acetic acid
SMILES OC(=O)Cc1ccc(OCC=C)c(Cl)c1

Specifications

Analyte or Component Names Alclofenac
Shipping Condition Room Temperature
InChI Formula InChI=1S/C11H11ClO3/c1-2-5-15-10-4-3-8(6-9(10)12)7-11(13)14/h2-4,6H,1,5,7H2,(H,13,14)
Solution Type Neat
Industry Type Forensics
Quantity 10 mg
Formula Weight 226.0397
Chemical Name or Material Alclofenac
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.